Learn More
GA International Dual Point Cryogenic Waterproof Permanent Marker Green 6/pack
Catalog No. NC2394831
Encompass_Preferred
Waterproof, cryogenic permanent marker with dual point, fine and extra-fine high-quality tips. This 2-in-1 marker is available in a range of colors, and has excellent performance on plastic, glass, metal, and cloth surfaces. Ideal for use in bio-medical laboratories, this permanent marker can be used to write on cryogenic labels, for subsequent storage in liquid nitrogen, and lab freezers.
Catalog No. NC2394831 Supplier GA International Supplier No. MP1G6
Waterproof, cryogenic permanent marker with dual point, fine and extra-fine high-quality tips. This 2-in-1 marker is available in a range of colors, and has excellent performance on plastic, glass, metal, and cloth surfaces. Ideal for use in bio-medical laboratories, this permanent marker can be used to write on cryogenic labels, for subsequent storage in liquid nitrogen, and lab freezers.