Learn More
GA International DYMO-Compatible Cryogenic Labels 0.5" x 1.04" + 0.375" (circle) / 12.7mm x 26.4mm + 9.5mm (circle) Rectangle + Circle Blue From -196°C to +70°C (-320.8°F to +158°F)
Catalog No. NC1745464
Encompass_Preferred
DYMO compatible direct thermal cryo labels. For ultra-low and cryogenic temperature applications (as low as -196°C). Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (-80°C, -50°C, -20°C).
Catalog No. NC1745464 Supplier GA International Supplier No. EG1F072BL
DYMO compatible direct thermal cryo labels. For ultra-low and cryogenic temperature applications (as low as -196°C). Ideal for identification and permanent storage of vials and containers in liquid nitrogen and laboratory freezers (-80°C, -50°C, -20°C).