Learn More
Aviva Systems Biology Corp CRYM Antibody - N-terminal region (ARP51981_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51981_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRYM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRYM. Alternate names: DFNA40, THBPApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-53982