Learn More
Aviva Systems Biology Corp CRYM Antibody - middle region (ARP51982_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51982_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRYM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CRYM. Alternate names: DFNA40, THBPApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53983