Learn More
Andwin Scientific Cryomold Biopsy 10X10X5mm 100/pk
SDP
Catalog No. NC9806558
Encompass_Preferred
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 10 mm x 10 mm x 5 mm. It is sold in a pack of 100.
Catalog No. NC9806558 Supplier Andwin Scientific Supplier No. 4565
The Cryomold molds produce uniformly shaped, flat-surface specimen block when used with O.C.T. (optimum cutting temperature) compound, which is sold separately, for frozen sectioning. This can fit into Cryobar freezing wells of the Tissue Tek II microtome/cryostat with the use of an adapter. This product measures 10 mm x 10 mm x 5 mm. It is sold in a pack of 100.