Learn More
Med Vet International Jorvet Cryogun Attachments, Opeing Site Aperature 0.04in., J0810AA
Catalog No. 5000495041
Encompass_Preferred
Straight Spray Attachments. NOTE: Open spray attachments are used for lesions on patient's outside body. The bigger the aperture, the larger the volume of spray dispensed.
Catalog No. 50-004-95041 Supplier Med Vet International Supplier No. JOR810AA
Straight Spray Attachments. NOTE: Open spray attachments are used for lesions on patient's outside body. The bigger the aperture, the larger the volume of spray dispensed.