Learn More
Andwin Scientific CRYO-BABIES 1.28 L X 0.5 W
SDP
Catalog No. 50821032
Encompass_Preferred
The Cryo-Babies are resistant against chemicals and are flexible. The labels can endure extreme freezer and autoclaving temperatures, water baths, and boiling without peeling, cracking, or degrading. These can also stick onto several surfaces, including plastics, glass, and metal. This product measures 1.28" x 0.5" and is sold in a pack of 1000.
Catalog No. 50-821-032 Supplier Andwin Scientific Supplier No. 91882000
The Cryo-Babies are resistant against chemicals and are flexible. The labels can endure extreme freezer and autoclaving temperatures, water baths, and boiling without peeling, cracking, or degrading. These can also stick onto several surfaces, including plastics, glass, and metal. This product measures 1.28" x 0.5" and is sold in a pack of 1000.