Learn More
USA Scientific Inc Cryo-Babies, 1.28 inch x 0.5 inch, green, on rolls
Catalog No. NC1662511
Encompass
Cryo-Babies, large, green labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. NC1662511 Supplier USA Scientific Inc Supplier No. 91882002
Cryo-Babies, large, green labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.