Learn More
USA Scientific Inc Cryo-Babies, 1.28 inch x 0.5 inch, blue, on rolls
Catalog No. NC1662510
Encompass
Cryo-Babies, large, blue labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. NC1662510 Supplier USA Scientific Inc Supplier No. 91882001
Cryo-Babies, large, blue labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.