Learn More
USA Scientific Inc Cryo-Babies, 1.28 inch x 0.5 inch, yellow, on rolls
Catalog No. NC0360238
Encompass
Cryo-Babies yellow labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. NC0360238 Supplier USA Scientific Inc Supplier No. 91882006
Cryo-Babies yellow labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.