Learn More
Andwin Scientific Cryo-Babies, small, white labels for 0.5 ml tubes, 0.94 in. L x 0.5 in. W, 1000/roll. For -196C to +150C., EA
SDP
Catalog No. NC9501464
Encompass_Preferred
Cryo-Babies, small, white labels for 0.5 ml tubes, 0.94 in. L x 0.5 in. W, 1000/roll. For -196C to +150C., EA
Catalog No. NC9501464 Supplier Andwin Scientific Supplier No. 91880500
Cryo-Babies, small, white labels for 0.5 ml tubes, 0.94 in. L x 0.5 in. W, 1000/roll. For -196C to +150C., EA