Learn More
Sigma Aldrich Fine Chemicals Biosciences Cryo-BabiesR Labels white
Catalog No. 502900789
Encompass
Pre-cut chemically inert vinyl labels withstand boiling water conventional freezing and liquid nitrogen storage without peeling or cracking. Labels will accept any marking instrument and easily peel off backing sheet for application to vial. Size is 1.28 x 0.5 in. fitting precisely around standard cryogenic vials as well as 1.5 to 2 mL microcentrifuge tubes. Cryo-Babies also withstand flash/snap freezing in liquid nitrogen boiling water baths at 100 C and dry heat up to 150 C.
Catalog No. 50-290-0789 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3662181PAK
Pre-cut chemically inert vinyl labels withstand boiling water conventional freezing and liquid nitrogen storage without peeling or cracking. Labels will accept any marking instrument and easily peel off backing sheet for application to vial. Size is 1.28 x 0.5 in. fitting precisely around standard cryogenic vials as well as 1.5 to 2 mL microcentrifuge tubes. Cryo-Babies also withstand flash/snap freezing in liquid nitrogen boiling water baths at 100 C and dry heat up to 150 C.