Learn More
Med Vet International Jorvet Cryosurgery Black Vent Adapter, J0810J
Catalog No. 5000495218
Encompass_Preferred
Eliminates intermittent spray and increases nitrogen flow when used with small contact probes and smaller apertures.
Catalog No. 50-004-95218 Supplier Med Vet International Supplier No. JOR810J
Eliminates intermittent spray and increases nitrogen flow when used with small contact probes and smaller apertures.