Learn More
Stellar Scientific Cardboard Cryo Freezer Box for 50mL Tubes, 16 Places with Divider, 48/cs
SDP
Catalog No. NC1834841
Encompass
These sturdy cardboard boxes are specially coated to resist softening from repeated freeze and thaw cycles. Smart looking with a well sealing lid and a generous amount of room to write on top; we get heaps of praises about these.
Catalog No. NC1834841 Supplier Stellar Scientific Supplier No. STO05048
These sturdy cardboard boxes are specially coated to resist softening from repeated freeze and thaw cycles. Smart looking with a well sealing lid and a generous amount of room to write on top; we get heaps of praises about these.