Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Cryo Box Holder
SDP

Catalog No. 22000676
Change view
Click to view available options
Holds:
14 Boxes
28 Boxes
Mounting Type:
Magnet
Wall Mount
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Mounting Type
22-000-676 14 Boxes Magnet
22-000-677 14 Boxes Wall Mount
22-000-678 28 Boxes Magnet
22-000-679 28 Boxes Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-000-676 Supplier Poltex Supplier No. CRYO14M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Poltex Cryo Box Holder includes a pocket in front for pens, implements, tape.

  • Holds stacks of 5 x 5 x 2 in. cryo boxes.
  • Pocket in front for pens, implements, tape.
  • Durable PETG construction.
  • Choose from wall mount (screws/anchors included), or magnet mount (powerful magnets included).
    • Wall mount version either mounts to wall with screws/keyhold slots, or stands on counter (comes with non-slip rubber bumpons on base).
  • Choose from 14 or 28 box capacity.
  • See image for dimensions

Specifications

Color Transparent
For Use With (Equipment) Cryo Boxes
Holds 14 Boxes
Material Polycarbonate
Type Cryogenic Box Storage Rack
Mounting Type Magnet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.