Learn More
Genesee Scientific Corporation Genesee Scientific 27-337, 81-Place Cryogenic Box with Lid, Blue, Holds 1.2 - 2ml Tubes, 5 Boxes/Unit
SDP
Supplier:  Genesee Scientific Corporation 27337B
Encompass
2-inch assorted color polycarbonate box with an 81-place gridMade of durable polycarbonate that withstands temperatures between -196°C to +121°CComes in bright, attractive assorted colors of blue, green, orange, red and yellowIndex numerals printed on the rack base for fast sample identificationRacks readily accommodate all standard 1.2 - 2ml microtubes and cryovialsVents and holes in both base and lid for drainingDimensions (W x D x H): 5.25 x 5.25 x 2.125in / 133 x 133 x 54mm
Catalog No. NC3065544