Learn More
Med Vet International Jorvet Cryogun Attachments, 10mm Cryochamber, J0810DB
Catalog No. 5000495816
Encompass_Preferred
Permits precise confined spray. Liquid nitrogen pools over deep lesions. Suggested use with malleable extensions (e.g., JOR810ra)
Catalog No. 50-004-95816 Supplier Med Vet International Supplier No. JOR810DB
Permits precise confined spray. Liquid nitrogen pools over deep lesions. Suggested use with malleable extensions (e.g., JOR810ra)