Learn More
Med Vet International Jorvet Cryogun Attachments, 15mm Cryochamber, J0810DC
Catalog No. 5000495842
Encompass_Preferred
Permits precise confined spray. Liquid nitrogen pools over deep lesions. Suggested use with malleable extensions (e.g., JOR810ra)
Catalog No. 50-004-95842 Supplier Med Vet International Supplier No. JOR810DC
Permits precise confined spray. Liquid nitrogen pools over deep lesions. Suggested use with malleable extensions (e.g., JOR810ra)