Learn More
Research Products International Corp Cryo Clear Laser Labels for 0.5 ml Tubes, 2380 per Package
SDP
Catalog No. 50998650
Encompass
Cryo-Clear laser printable labels withstand cryogenic storage conditions at -196°C and high temperatures up to +70°C. Chemically inert labels resist most organic solvents and adhere to plastic, glass and metal without cracking, peeling or degrading. Label sheets are specially coated to accept laser printing and other methods of marking. Not recommended for use with siliconized tubes.
Catalog No. 50-998-650 Supplier Research Products International Corp Supplier No. 247169
Add to Cart
Add to Cart
Cryo-Clear laser printable labels withstand cryogenic storage conditions at -196°C and high temperatures up to +70°C. Chemically inert labels resist most organic solvents and adhere to plastic, glass and metal without cracking, peeling or degrading. Label sheets are specially coated to accept laser printing and other methods of marking. Not recommended for use with siliconized tubes.