Learn More
Med Vet International Jorvet Cryogun Attachments, 3mm Conical Shaped Closed Probe, J0810EC
Catalog No. 5000495845
Encompass_Preferred
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.
Catalog No. 50-004-95845 Supplier Med Vet International Supplier No. JOR810EC
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.