Learn More
Med Vet International Jorvet Cryogun Attachments, 4mm Conical-Shaped Closed Probe, J0810ED
Catalog No. 5000495868
Encompass_Preferred
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.
Catalog No. 50-004-95868 Supplier Med Vet International Supplier No. JOR810ED
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.