Learn More
Med Vet International Jorvet Cryogun Attachments, 5mm Conical-Shaped Closed Probe, J0810EF
Catalog No. 5000495837
Encompass_Preferred
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.
Catalog No. 50-004-95837 Supplier Med Vet International Supplier No. JOR810EF
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.