Learn More
Med Vet International Jorvet Cryogun Attachments, 6mm Conical-Shaped Closed Probe, J0810EG
Catalog No. 5000495919
Encompass_Preferred
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.
Catalog No. 50-004-95919 Supplier Med Vet International Supplier No. JOR810EG
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.