Learn More
Med Vet International Jorvet Cryosurgery Cryoplate, J0810M
Catalog No. 5000495280
Encompass_Preferred
A transparent lexan plate with four various size conical openings. Used to concentrate spray within a limited area.
Catalog No. 50-004-95280 Supplier Med Vet International Supplier No. JOR810M
A transparent lexan plate with four various size conical openings. Used to concentrate spray within a limited area.