Learn More
GA International Cryogenic Color Dots for Tops of Microtubes 0.5" / 12.7mm Circle White From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC2166677
Encompass_Preferred
These small diameter round cryo labels for microtube tops* are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. LT-series circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.
This item is not returnable. View return policy
Catalog No. NC2166677 Supplier GA International Supplier No. LT13WH
These small diameter round cryo labels for microtube tops* are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. LT-series circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.