Learn More
GA International Cryogenic Color Dots for Tops of 0.5 mL & 1.5 mL Microtubes 0.354" / 9mm Circle Pink From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC1892819
Encompass_Preferred
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. Specially designed to comfortably fit 0.5 ml, 0.6 ml, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.
Catalog No. NC1892819 Supplier GA International Supplier No. LT9PI
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage. Specially designed to comfortably fit 0.5 ml, 0.6 ml, and 1.5 ml Eppendorf tube caps and microtubes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.