Learn More
Research Products International Corp Cryo Dual-Point Markers, Asst, 8/pk
SDP
Catalog No. 502252978
Encompass
Water resistant Cryo Dual-Point Markers offer a fine (0.7mm) and extra fine (0.3mm) tip. Permanent markers offer excellent performance for use in various laboratory applications including cryogenic labels, liquid nitrogen storage, and lab freezers. Provide a marking on most surfaces (plastic, glass, metal, cloth, Teflon) and withstand liquid nitrogen temperatures down to -196°C. Available as single color or assorted (black, brown, green, blue, purple, red, orange and yellow).
Water resistant
Permanent ink
High Quality tip
This item is not returnable. View return policy
Catalog No. 50-225-2978 Supplier Research Products International Corp Supplier No. 248511
Water resistant Cryo Dual-Point Markers offer a fine (0.7mm) and extra fine (0.3mm) tip. Permanent markers offer excellent performance for use in various laboratory applications including cryogenic labels, liquid nitrogen storage, and lab freezers. Provide a marking on most surfaces (plastic, glass, metal, cloth, Teflon) and withstand liquid nitrogen temperatures down to -196°C. Available as single color or assorted (black, brown, green, blue, purple, red, orange and yellow).
Water resistant
Permanent ink
High Quality tip