Learn More
Mitegen Advanced Cryo-EM Grid Storage & Shipping System, Incl. Pucks, Canes, Tongs, Dewar, Tools, For Organized Sample Workflow
SDP
Catalog No. NC1863740
Encompass_Preferred
The Advanced Cryo-EM Grid Storage and Shipping System is a comprehensive kit designed to organize, store, track, and safely ship your valuable samples on grids for single particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications. It includes 10 2nd Generation Cryo-EM Pucks (M-CEMG2-10), offering 2D barcoding and custom serialization for precise tracking, along with a choice of 10 unique puck colors for organization. The kit features one Storage Cane (M-CEMSTC-2) compatible with TW34, HC34, TW35-6, VHC35 dewars, and one Shipping Cane (M-CEMTPC-1) compatible with CX100 dryshippers. Essential handling tools are included: Puck Grasping Tongs (M-CEMG2-PGT1), Angled Cryo Tongs (M-CP-111-030), a Cryo-EM Puck Dewar with Insert (M-CEM-PD-IS1), Cryo-EM Grid Box Tweezers (TW-EMTEC-553), and a Screwdriver for Puck Lid Lock (M-CEMG2-LLSD). Log sheets (M-CEMG2-LogSheets) further aid tracking.
Catalog No. NC1863740 Supplier Mitegen Supplier No. MCEMG2ALSK1
The Advanced Cryo-EM Grid Storage and Shipping System is a comprehensive kit designed to organize, store, track, and safely ship your valuable samples on grids for single particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications. It includes 10 2nd Generation Cryo-EM Pucks (M-CEMG2-10), offering 2D barcoding and custom serialization for precise tracking, along with a choice of 10 unique puck colors for organization. The kit features one Storage Cane (M-CEMSTC-2) compatible with TW34, HC34, TW35-6, VHC35 dewars, and one Shipping Cane (M-CEMTPC-1) compatible with CX100 dryshippers. Essential handling tools are included: Puck Grasping Tongs (M-CEMG2-PGT1), Angled Cryo Tongs (M-CP-111-030), a Cryo-EM Puck Dewar with Insert (M-CEM-PD-IS1), Cryo-EM Grid Box Tweezers (TW-EMTEC-553), and a Screwdriver for Puck Lid Lock (M-CEMG2-LLSD). Log sheets (M-CEMG2-LogSheets) further aid tracking.