Learn More
Mitegen Cryo Grid Box for Clipped (Autogrid) Grids, Holds 4 AutoGrids, Screw-On Lid for Secure Cryo Transfers, Sample Protection
SDP
Catalog No. NC1863758
Encompass
This Cryo Grid Box is expertly designed for secure storage and handling of clipped grids, often referred to as autogrids, in cryo-electron microscopy (cryo-EM) workflows. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1. Each robust grid box is capable of holding up to four AutoGrids, ensuring efficient organization and protection of your valuable samples. It features a reliable screw-on lid that firmly holds the AutoGrids in place, safeguarding them from accidental dislodgement and physical damage during cryo transfers. This secure design is essential for maintaining the integrity of delicate samples in cryogenic environments. Also commonly known as an AutoGrid box, this product streamlines your cryo-sample management, making it an indispensable tool for laboratories conducting high-resolution cryo-EM research where sample protection is paramount during handling and storage.
Catalog No. NC1863758 Supplier Mitegen Supplier No. MCEM7AGB
This Cryo Grid Box is expertly designed for secure storage and handling of clipped grids, often referred to as autogrids, in cryo-electron microscopy (cryo-EM) workflows. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1. Each robust grid box is capable of holding up to four AutoGrids, ensuring efficient organization and protection of your valuable samples. It features a reliable screw-on lid that firmly holds the AutoGrids in place, safeguarding them from accidental dislodgement and physical damage during cryo transfers. This secure design is essential for maintaining the integrity of delicate samples in cryogenic environments. Also commonly known as an AutoGrid box, this product streamlines your cryo-sample management, making it an indispensable tool for laboratories conducting high-resolution cryo-EM research where sample protection is paramount during handling and storage.