Learn More
Mitegen Cryo-EM AutoGrid Cassette Puck for Glacios and Mitegen Storage and Transport Cryo-EM Puck Canes
SDP
Catalog No. 5000495006
Encompass_Preferred
The MiTeGen AutoGrid Cassette Storage and Shipping Puck is based on the MiTeGen 2nd generation Cryo-EM Puck. This puck is designed to allow users to screen on a Glacios and then ship the same AutoGrid cassette puck to a collection facility, or for facilities to store their cassettes in puck canes. The puck is also designed to fit into a cassette loading station. It works with the following storage, shipping canes, and tools: ICB34, ICB35-6, HC34, and VHC35 storage canes (M-CEMSTC-2), TW35-10, and HC35 storage canes (M-CEMSTC-2-35), Puck shipping cane (M-CEMTPC-1), and Grasping tongs (M-CEMG2-PGT1). The puck can be barcoded or engraved for tracking purposes.
Catalog No. 50-004-95006 Supplier Mitegen Supplier No. MCEMAGCP1
The MiTeGen AutoGrid Cassette Storage and Shipping Puck is based on the MiTeGen 2nd generation Cryo-EM Puck. This puck is designed to allow users to screen on a Glacios and then ship the same AutoGrid cassette puck to a collection facility, or for facilities to store their cassettes in puck canes. The puck is also designed to fit into a cassette loading station. It works with the following storage, shipping canes, and tools: ICB34, ICB35-6, HC34, and VHC35 storage canes (M-CEMSTC-2), TW35-10, and HC35 storage canes (M-CEMSTC-2-35), Puck shipping cane (M-CEMTPC-1), and Grasping tongs (M-CEMG2-PGT1). The puck can be barcoded or engraved for tracking purposes.