Learn More
Mitegen Barcoded Cryo Grid Box, Autogrid Compatible, 2D Barcode for Easy Sample Tracking, For Cryo-EM Sample Storage
SDP
Catalog No. NC1863754
Encompass
This Barcoded Cryo Grid Box offers an advanced solution for the secure storage and precise tracking of clipped grids, commonly known as autogrids, within cryo-electron microscopy (cryo-EM) workflows. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1. Each robust grid box is designed for compatibility with autogrid formats and features an integrated 2D barcode. This unique identification system enables seamless and accurate sample tracking throughout your experimental process, from preparation to data acquisition. The barcoding minimizes manual logging errors and enhances overall laboratory efficiency, crucial for high-throughput research. This grid box provides reliable protection for delicate samples in cryogenic environments, safeguarding them from damage during handling and storage.
Catalog No. NC1863754 Supplier Mitegen Supplier No. MCEM7AGBBC1
This Barcoded Cryo Grid Box offers an advanced solution for the secure storage and precise tracking of clipped grids, commonly known as autogrids, within cryo-electron microscopy (cryo-EM) workflows. This grid box can be used in the MiTeGen Generation 2 Cryo-EM puck SKU M-CEMG2-1. Each robust grid box is designed for compatibility with autogrid formats and features an integrated 2D barcode. This unique identification system enables seamless and accurate sample tracking throughout your experimental process, from preparation to data acquisition. The barcoding minimizes manual logging errors and enhances overall laboratory efficiency, crucial for high-throughput research. This grid box provides reliable protection for delicate samples in cryogenic environments, safeguarding them from damage during handling and storage.