Learn More
Mitegen Basic Cryo-EM Grid Storage & Shipping System, Incl. 10 Pucks, Canes, Tongs, For Organized Sample Workflow
SDP
Catalog No. NC1863742
Encompass_Preferred
This Basic Cryo-EM Grid Storage and Shipping System is a comprehensive kit designed to organize, store, track, and safely ship your valuable samples on grids for single particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency. The kit includes 10 2nd Generation Cryo-EM Pucks (M-CEMG2-10) with free barcoding and serialization for precise tracking, along with a free puck color selection from 10 unique colors for organization. It features one Storage Cane (M-CEMSTC-2) compatible with TW34, HC34, TW35-6, VHC35 dewars, and one Shipping Cane (M-CEMTPC-1) compatible with CX100 dryshippers. A Puck Grasping Tong (M-CEMG2-PGT1) is also included for easy handling. This system provides an excellent foundational solution for managing cryo-EM samples, streamlining storage, tracking, and transport processes for high-resolution research.
Catalog No. NC1863742 Supplier Mitegen Supplier No. MCEMG2BLSK1
This Basic Cryo-EM Grid Storage and Shipping System is a comprehensive kit designed to organize, store, track, and safely ship your valuable samples on grids for single particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency. The kit includes 10 2nd Generation Cryo-EM Pucks (M-CEMG2-10) with free barcoding and serialization for precise tracking, along with a free puck color selection from 10 unique colors for organization. It features one Storage Cane (M-CEMSTC-2) compatible with TW34, HC34, TW35-6, VHC35 dewars, and one Shipping Cane (M-CEMTPC-1) compatible with CX100 dryshippers. A Puck Grasping Tong (M-CEMG2-PGT1) is also included for easy handling. This system provides an excellent foundational solution for managing cryo-EM samples, streamlining storage, tracking, and transport processes for high-resolution research.