Learn More
Mitegen Cryo-EM Clipping Station with Foam Dewar and Clipping Hub By Nanosoft
SDP
Catalog No. 5000494989
Encompass
The Nanosoft Clipping Station provides ease of use & efficiency for the clipping of cryo-EM grids for Thermo Fisher autoloader-based microscopes. Its design is optimized for a seamless clipping workflow, & its high-quality foam dewar integrates grid box storage puck access directly adjacent to the clipping space. A new clipping design makes the workflow easier, & a well-designed lid balances ease of use & protection. The Clipping Station is compatible with both NanoSoft & Thermo Fisher clipping tools. Consists of a Clipping Hub, Clipping Station Lid, a Foam Dewar, Clipping Tool Puck Rack, Lid Plug and Base Mat.
Catalog No. 50-004-94989 Supplier Mitegen Supplier No. MCEMNSCLSTN01
The Nanosoft Clipping Station provides ease of use & efficiency for the clipping of cryo-EM grids for Thermo Fisher autoloader-based microscopes. Its design is optimized for a seamless clipping workflow, & its high-quality foam dewar integrates grid box storage puck access directly adjacent to the clipping space. A new clipping design makes the workflow easier, & a well-designed lid balances ease of use & protection. The Clipping Station is compatible with both NanoSoft & Thermo Fisher clipping tools. Consists of a Clipping Hub, Clipping Station Lid, a Foam Dewar, Clipping Tool Puck Rack, Lid Plug and Base Mat.