Learn More
Mitegen Shelved Transport Cane for MiTeGen Generation 2 Cryo-EM Pucks, Holds 8 Pucks, Designed for use with CX100 Dryshippers, Secure Sample Transport
SDP
Catalog No. NC1863753
Encompass_Preferred
This Shelved Transport Cane is meticulously designed for the safe and organized shipping of Cryo-EM Pucks in CX100 Dryshippers. Engineered for reliable performance during transit, this cane provides a secure solution for valuable cryogenic samples. It is specifically designed to accommodate Cryo-EM Pucks, including the MiTeGen 2nd generation cryo-EM puck SKU M-CEMG2-1, ensuring compatibility and stable positioning during transport. The cane is capable of holding up to eight pucks, optimizing space within the dryshipper and streamlining sample management for high-throughput cryo-electron microscopy workflows. Its robust construction minimizes exposure and maintains sample integrity by safeguarding specimens in ultra-low temperature environments throughout the shipping process. This shelved transport cane is an indispensable accessory for laboratories requiring dependable and compliant methods for moving cryo-EM samples between locations.
Catalog No. NC1863753 Supplier Mitegen Supplier No. MCEMTPC1
This Shelved Transport Cane is meticulously designed for the safe and organized shipping of Cryo-EM Pucks in CX100 Dryshippers. Engineered for reliable performance during transit, this cane provides a secure solution for valuable cryogenic samples. It is specifically designed to accommodate Cryo-EM Pucks, including the MiTeGen 2nd generation cryo-EM puck SKU M-CEMG2-1, ensuring compatibility and stable positioning during transport. The cane is capable of holding up to eight pucks, optimizing space within the dryshipper and streamlining sample management for high-throughput cryo-electron microscopy workflows. Its robust construction minimizes exposure and maintains sample integrity by safeguarding specimens in ultra-low temperature environments throughout the shipping process. This shelved transport cane is an indispensable accessory for laboratories requiring dependable and compliant methods for moving cryo-EM samples between locations.