Learn More
Mitegen Round Cryo Grid Box with Lid for Cryo-EM Sample Storage Pucks, Four Diamond Slots, Non-Static Cover, Stainless Steel Screw, Blue Color, 1 Each
SDP
Catalog No. NC1866179
Encompass
The round Cryo Grid Box with Lid is specifically engineered for secure and organized storage of cryo-EM samples within cryoem grid box storage pucks such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1. This robust cryo grid box is designed to protect delicate samples during handling and long-term storage, making it an essential component for high-resolution electron microscopy workflows. It features 4 precisely shaped diamond slots, ensuring stable and reliable placement of grids. The non-static cover, which effectively prevents electrostatic discharge that could damage sensitive samples, is securely held in place by a durable stainless steel screw tapped centrally into the box. Its distinct blue color aids in easy identification and organization within busy laboratory settings.
Catalog No. NC1866179 Supplier Mitegen Supplier No. MCEM7116610
The round Cryo Grid Box with Lid is specifically engineered for secure and organized storage of cryo-EM samples within cryoem grid box storage pucks such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1. This robust cryo grid box is designed to protect delicate samples during handling and long-term storage, making it an essential component for high-resolution electron microscopy workflows. It features 4 precisely shaped diamond slots, ensuring stable and reliable placement of grids. The non-static cover, which effectively prevents electrostatic discharge that could damage sensitive samples, is securely held in place by a durable stainless steel screw tapped centrally into the box. Its distinct blue color aids in easy identification and organization within busy laboratory settings.