Learn More
Mitegen Round Cryo Grid Box With Lid Designed For Use With Cryo-em Grid Box Storage Pucks - White In Color
SDP
Catalog No. NC1866180
Encompass
The round cryo grid box round with lid designed for use within cryoem grid box storage pucks such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1. Features four diamond shaped slots, non-static cover is held in place with stainless steel screw which is tapped in the center of the box. White in color.
Catalog No. NC1866180 Supplier Mitegen Supplier No. MCEM7116610W
The round cryo grid box round with lid designed for use within cryoem grid box storage pucks such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1. Features four diamond shaped slots, non-static cover is held in place with stainless steel screw which is tapped in the center of the box. White in color.