Learn More
Mitegen Round Cryo Grid Box with Pin Type Lid, For FEI Vitrobot Systems, Ensures Secure Grid & Sample Handling
SDP
Catalog No. NC1866186
Encompass
This Round Cryo Grid Box with Pin Type Lid is specifically designed to provide practical and secure grid storage in cryogenic conditions, optimized for seamless integration with FEI Vitrobot systems. The innovative pin type lid ensures a firm and precise closure, actively safeguarding delicate cryo-EM grids and samples from damage or contamination during handling and transfer. Its robust construction supports reliable performance in demanding laboratory environments. Ideal for cryo-electron microscopy workflows, this grid box facilitates efficient and organized sample management, ensuring that valuable specimens are protected at ultra-low temperatures. The design promotes easy grid insertion and retrieval, streamlining the vitrification process and subsequent transfer for high-resolution imaging. This single unit offers a dependable solution for microscopists requiring specialized tools for their FEI Vitrobot setup.
Catalog No. NC1866186 Supplier Mitegen Supplier No. MCEM7116640
This Round Cryo Grid Box with Pin Type Lid is specifically designed to provide practical and secure grid storage in cryogenic conditions, optimized for seamless integration with FEI Vitrobot systems. The innovative pin type lid ensures a firm and precise closure, actively safeguarding delicate cryo-EM grids and samples from damage or contamination during handling and transfer. Its robust construction supports reliable performance in demanding laboratory environments. Ideal for cryo-electron microscopy workflows, this grid box facilitates efficient and organized sample management, ensuring that valuable specimens are protected at ultra-low temperatures. The design promotes easy grid insertion and retrieval, streamlining the vitrification process and subsequent transfer for high-resolution imaging. This single unit offers a dependable solution for microscopists requiring specialized tools for their FEI Vitrobot setup.