Learn More
Mitegen Cryo-EM Grid Box Tweezers, Easy-to-Use Design, Grips Central Screw for Secure Handling & Sample Protection
SDP
Catalog No. NC1446557
Encompass
These Cryo-EM Grid Box Tweezers are ingeniously designed to simplify the handling of delicate cryo grid boxes, making your cryo-electron microscopy workflow more efficient and secure. Unlike conventional tweezers that grip the sides of the box, this specialized tool is meticulously crafted to securely grip the box's central screw. This unique design ensures a firm, stable hold, preventing accidental slips or drops during critical transfers and manipulations, thereby safeguarding your valuable samples. The easy-to-use nature of these tweezers reduces the risk of damage to both the grid box and its precious contents. By providing enhanced control and stability, these tweezers are an indispensable accessory for any laboratory performing cryo-EM sample preparation, storage, and transfer, ensuring sample integrity and streamlining handling procedures in cryogenic environments. Compatible with every pin lid type grid box.
Catalog No. NC1446557 Supplier Mitegen Supplier No. TWEMTEC553
These Cryo-EM Grid Box Tweezers are ingeniously designed to simplify the handling of delicate cryo grid boxes, making your cryo-electron microscopy workflow more efficient and secure. Unlike conventional tweezers that grip the sides of the box, this specialized tool is meticulously crafted to securely grip the box's central screw. This unique design ensures a firm, stable hold, preventing accidental slips or drops during critical transfers and manipulations, thereby safeguarding your valuable samples. The easy-to-use nature of these tweezers reduces the risk of damage to both the grid box and its precious contents. By providing enhanced control and stability, these tweezers are an indispensable accessory for any laboratory performing cryo-EM sample preparation, storage, and transfer, ensuring sample integrity and streamlining handling procedures in cryogenic environments. Compatible with every pin lid type grid box.