Learn More
Molecular Dimensions Cryo-EM grid-boxes, 20/pk
SDP
Catalog No. NC1620771
Encompass_Preferred
This new grid-box from Swissci has been developed in collaboration with Dr Jan Löwe at the MRC Laboratory of Molecular Biology in Cambridge. Backed by years of experience in Cryo-EM, it includes many improvements on current models. The Swissci Cryo-EM Grid-Box is made from a special polymer formulation, which ensures there is no sample-damaging static charging at cryogenic temperatures. No screwdriver is required since the lid is rotated with standard laboratory tweezers. No need to hand label each box and then struggle to interpret the label. Each grid-box is provided with a pressure adhesive cryo-label with a unique identification number. Each diamond-shape slot is marked by a roman numeral. The Swissci cryo-EM grid-box is the standard size for all common cryo-EM sample mounting and storage workflows. Cost-effective pricing means the grid-box is truly disposable: avoid the confusion of multiple samples having the same identification number by using each grid-box a single time.
Catalog No. NC1620771 Supplier Molecular Dimensions Supplier No. MD16104
This new grid-box from Swissci has been developed in collaboration with Dr Jan Löwe at the MRC Laboratory of Molecular Biology in Cambridge. Backed by years of experience in Cryo-EM, it includes many improvements on current models. The Swissci Cryo-EM Grid-Box is made from a special polymer formulation, which ensures there is no sample-damaging static charging at cryogenic temperatures. No screwdriver is required since the lid is rotated with standard laboratory tweezers. No need to hand label each box and then struggle to interpret the label. Each grid-box is provided with a pressure adhesive cryo-label with a unique identification number. Each diamond-shape slot is marked by a roman numeral. The Swissci cryo-EM grid-box is the standard size for all common cryo-EM sample mounting and storage workflows. Cost-effective pricing means the grid-box is truly disposable: avoid the confusion of multiple samples having the same identification number by using each grid-box a single time.