Learn More
Mitegen Cryo-EM Grid Storage and Shipping Puck Storage Case - Hard Plastic Case with Handle
SDP
Catalog No. 5000495015
Encompass_Preferred
The MiTeGen Cryo-EM grid storage and shipping puck storage case is a durable case designed to hold the tools found in either the Cryo-EM Basic Grid Storage and Shipping Puck System INSERT SKU or the Cryo-EM Advanced Grid Storage and Shipping Puck System INSERT SKU. The case is made of a hard plastic and features a carrying handle, making it easy to transport your tools. Holds 1 Storage Cane and 10 pucks, 1 Transport Cane, Puck Grasping Tongs, Bent Cryo Tongs, and extra space for grid boxes and other tools.
Catalog No. 50-004-95015 Supplier Mitegen Supplier No. MCEMG2SC1
The MiTeGen Cryo-EM grid storage and shipping puck storage case is a durable case designed to hold the tools found in either the Cryo-EM Basic Grid Storage and Shipping Puck System INSERT SKU or the Cryo-EM Advanced Grid Storage and Shipping Puck System INSERT SKU. The case is made of a hard plastic and features a carrying handle, making it easy to transport your tools. Holds 1 Storage Cane and 10 pucks, 1 Transport Cane, Puck Grasping Tongs, Bent Cryo Tongs, and extra space for grid boxes and other tools.