Learn More
Mitegen Cryo-EM ICB34 Facility Grid Storage and Shipping Puck System for Labs - Includes 60 Pucks, 6 Storage Canes, 1 Shipping Cane, Tongs, Dewar, Barcode Reader, and more
SDP
Catalog No. 5000495035
Encompass
The MiTeGen Cryo-EM ICB34 Facility Grid Storage and Shipping System is designed for labs with multiple researchers, groups, or projects that need to store a large number of samples. This comprehensive kit, which enables you to organize, store, track, and ship samples on grids for single-particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency, includes 60 Cryo-EM pucks (M-CEMG2-1), six shelved storage canes (M-CEMSTC-2), one shelved transport cane (M-CEMTPC-1), one puck grasping tongs (M-CEMG2-PGT1), one angled cryo tongs for puck manipulation (M-CP-111-030), one Cryo-EM puck dewar and insert (M-CEM-PD-IS1), one Cryo-EM grid box tweezers (TW-EMTEC-553), one screwdriver for puck lid lock (M-CEMG2-LLSD), one USB barcode reader (M-BCR-001), one set of Cryo-EM puck and storage cane log sheets (M-CEMG2-LogSheets), and free puck color selection. Free puck barcoding and engraving are also included.
Catalog No. 50-004-95035 Supplier Mitegen Supplier No. MCEMG2FSK134
The MiTeGen Cryo-EM ICB34 Facility Grid Storage and Shipping System is designed for labs with multiple researchers, groups, or projects that need to store a large number of samples. This comprehensive kit, which enables you to organize, store, track, and ship samples on grids for single-particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency, includes 60 Cryo-EM pucks (M-CEMG2-1), six shelved storage canes (M-CEMSTC-2), one shelved transport cane (M-CEMTPC-1), one puck grasping tongs (M-CEMG2-PGT1), one angled cryo tongs for puck manipulation (M-CP-111-030), one Cryo-EM puck dewar and insert (M-CEM-PD-IS1), one Cryo-EM grid box tweezers (TW-EMTEC-553), one screwdriver for puck lid lock (M-CEMG2-LLSD), one USB barcode reader (M-BCR-001), one set of Cryo-EM puck and storage cane log sheets (M-CEMG2-LogSheets), and free puck color selection. Free puck barcoding and engraving are also included.