Learn More
Mitegen Cryo-EM ICB35 Facility Grid Storage and Shipping Puck System for Large Labs - Includes 100 Pucks, 10 Storage Canes, Shipping Cane, and More
SDP
Catalog No. 5000495014
Encompass
The MiTeGen Cryo-EM ICB35 Facility Grid Storage and Shipping System is a comprehensive kit designed for labs and groups that need to store and manage a large number of samples. It enables you to organize, store, track, and ship your samples on grids for single-particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency. The system includes 100 Cryo-EM pucks (M-CEMG2-1), 10 ICB35 storage canes (M-CEMSTC-2-35), one shipping cane (M-CEMTPC-1), one puck grasping tongs (M-CEMG2-PGT1), one angled puck tongs (M-CP-111-030), one puck dewar and insert (M-CEM-PD-IS1), one set of Cryo-EM grid box tweezers (TW-EMTEC-553), one screwdriver for the puck lid lock (M-CEMG2-LLSD), one USB barcode reader (M-BCR-001), and puck tracking log sheets (M-CEMG2-LogSheets). This kit also comes with free puck barcoding, engraving, and color selection.
Catalog No. 50-004-95014 Supplier Mitegen Supplier No. MCEMG2FSK135
The MiTeGen Cryo-EM ICB35 Facility Grid Storage and Shipping System is a comprehensive kit designed for labs and groups that need to store and manage a large number of samples. It enables you to organize, store, track, and ship your samples on grids for single-particle analysis and other Cryogenic Electron Microscopy (Cryo-EM) applications with ease and efficiency. The system includes 100 Cryo-EM pucks (M-CEMG2-1), 10 ICB35 storage canes (M-CEMSTC-2-35), one shipping cane (M-CEMTPC-1), one puck grasping tongs (M-CEMG2-PGT1), one angled puck tongs (M-CP-111-030), one puck dewar and insert (M-CEM-PD-IS1), one set of Cryo-EM grid box tweezers (TW-EMTEC-553), one screwdriver for the puck lid lock (M-CEMG2-LLSD), one USB barcode reader (M-BCR-001), and puck tracking log sheets (M-CEMG2-LogSheets). This kit also comes with free puck barcoding, engraving, and color selection.