Learn More
Mitegen CXR100 Cryo Express Dry Shipper By IC Biomedical With Replaceable Adsorbent, Designed For Cryogenic Transport, IATA Compliant
SDP
Catalog No. NC2326617
Encompass
The CXR100 Cryo Express Dry Shipper is engineered for the safe transport of various biological materials at cryogenic temperatures, consistently maintaining approximately -190°C. Its unique adsorbent material securely holds liquid nitrogen, preventing spills even if the unit is tipped, ensuring highly dependable transit. A key feature of the CXR100 is its replaceable adsorbent material, enabling extended product lifespan and consistent, optimal performance over time. This dry shipper fully complies with IATA regulations for transporting non-infectious biological samples, offering peace of mind and simplified logistics. Built with new rugged construction, a dependable necktube, and superior vacuum insulation, it provides a static holding time of up to 16 days and a working time of 11 days. It adsorbs 3.7 liters of LN2, weighing 18.3 lbs when full.
Catalog No. NC2326617 Supplier Mitegen Supplier No. TWCXR100
The CXR100 Cryo Express Dry Shipper is engineered for the safe transport of various biological materials at cryogenic temperatures, consistently maintaining approximately -190°C. Its unique adsorbent material securely holds liquid nitrogen, preventing spills even if the unit is tipped, ensuring highly dependable transit. A key feature of the CXR100 is its replaceable adsorbent material, enabling extended product lifespan and consistent, optimal performance over time. This dry shipper fully complies with IATA regulations for transporting non-infectious biological samples, offering peace of mind and simplified logistics. Built with new rugged construction, a dependable necktube, and superior vacuum insulation, it provides a static holding time of up to 16 days and a working time of 11 days. It adsorbs 3.7 liters of LN2, weighing 18.3 lbs when full.