Learn More
Mitegen CXR500 Cryo Express Dry Shipper From IC Biomedical, Provides Cryogenic Transport, IATA Compliant
SDP
Catalog No. NC3372996
Encompass
CXR500 Cryo Express (CX) Dry Shipper from IC Biomedical is designed for the safe transport of various materials at cryogenic temperatures down to approximately -190°C. This dry shipper complies with IATA regulations for transporting non-infectious biological samples, ensuring smooth transit. Built with new rugged construction, including a dependable necktube, ribbed high-strength aluminum body, and durable paint system, the CXR500 offers superior vacuum performance for maximum holding times of up to 11 days. It boasts a liquid nitrogen adsorbed capacity of 7 liters and a working time of 7 days, weighing 42.5 lbs when full. The CXR-500 exclusively accommodates the Stainless Steel 100 Cell 5 Shelf Inventory Rack with Boxes for Vials (both sold separately); it is not designed for shipping canes or canisters. Overall dimensions are 26.9 inches height, 15.5 inches diameter, and an 8.5-inch neck diameter. It can hold up to 500 vials.
Catalog No. NC3372996 Supplier Mitegen Supplier No. TWCXR500
CXR500 Cryo Express (CX) Dry Shipper from IC Biomedical is designed for the safe transport of various materials at cryogenic temperatures down to approximately -190°C. This dry shipper complies with IATA regulations for transporting non-infectious biological samples, ensuring smooth transit. Built with new rugged construction, including a dependable necktube, ribbed high-strength aluminum body, and durable paint system, the CXR500 offers superior vacuum performance for maximum holding times of up to 11 days. It boasts a liquid nitrogen adsorbed capacity of 7 liters and a working time of 7 days, weighing 42.5 lbs when full. The CXR-500 exclusively accommodates the Stainless Steel 100 Cell 5 Shelf Inventory Rack with Boxes for Vials (both sold separately); it is not designed for shipping canes or canisters. Overall dimensions are 26.9 inches height, 15.5 inches diameter, and an 8.5-inch neck diameter. It can hold up to 500 vials.