Learn More
Mitegen CX100 Cryo Express Dry Shipper By IC Biomedical, Provides -190°C Cryogenic Transport, IATA Compliant
SDP
Catalog No. 501187353
Encompass
CX100 Cryo Express (CX) Dry Shipper from IC Biomedical provides safe transport of various biological materials at cryogenic temperatures down to approximately -190°C. It complies with IATA regulations for transporting non-infectious biological samples. It offers maximum holding times of up to 18 days. With a liquid nitrogen adsorbed capacity of 4.1 liters and a working time of 12.6 days, it weighs 19 lbs when full. Key dimensions are 18.4 inches height, 9.2 inches diameter, and a 2.78-inch neck diameter. Comes with Necktube core & one stainless steel canister. Canister dimensions are: 2.64" x 11". The canister can hold: 85 1.2ml & 2.0ml Vials (5/cane) or 102 1.2ml & 2.0ml Vials (6/cane) or 280 1/2 Straws (10/cane) or 800 1/2 cc Straws - Bulk (2 levels). To use the CX100 for shipping of UniPucks, ALS Pucks, or Cryo-EM Pucks - shipping canes are required. For Unipuck & ALS pucks see SKU M-CP-111-065. For the MiTeGen Generation 2 Cryo-EM pucks see SKU M-CEMTPC-1.
Catalog No. 50-118-7353 Supplier Mitegen Supplier No. TWCX100
CX100 Cryo Express (CX) Dry Shipper from IC Biomedical provides safe transport of various biological materials at cryogenic temperatures down to approximately -190°C. It complies with IATA regulations for transporting non-infectious biological samples. It offers maximum holding times of up to 18 days. With a liquid nitrogen adsorbed capacity of 4.1 liters and a working time of 12.6 days, it weighs 19 lbs when full. Key dimensions are 18.4 inches height, 9.2 inches diameter, and a 2.78-inch neck diameter. Comes with Necktube core & one stainless steel canister. Canister dimensions are: 2.64" x 11". The canister can hold: 85 1.2ml & 2.0ml Vials (5/cane) or 102 1.2ml & 2.0ml Vials (6/cane) or 280 1/2 Straws (10/cane) or 800 1/2 cc Straws - Bulk (2 levels). To use the CX100 for shipping of UniPucks, ALS Pucks, or Cryo-EM Pucks - shipping canes are required. For Unipuck & ALS pucks see SKU M-CP-111-065. For the MiTeGen Generation 2 Cryo-EM pucks see SKU M-CEMTPC-1.