Learn More
Med Vet International Jorvet Cryogun Attachments, 1cm Flat Probe, J0810FA
Catalog No. 5000495921
Encompass_Preferred
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).
Catalog No. 50-004-95921 Supplier Med Vet International Supplier No. JOR810FA
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).