Learn More
Med Vet International Jorvet Cryogun Attachments, 2.5cm Flat Probe, J0810FC
Catalog No. 5000495869
Encompass_Preferred
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).
Catalog No. 50-004-95869 Supplier Med Vet International Supplier No. JOR810FC
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).