Learn More
Med Vet International Jorvet Cryogun Attachments, 2cm Flat Probe, J0810FB
Catalog No. 5000495876
Encompass_Preferred
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).
Catalog No. 50-004-95876 Supplier Med Vet International Supplier No. JOR810FB
Circular, and useful for wide flat lesions (e.g., squamous cell carcinoma and sarcoids).