Learn More
Stellar Scientific Cardboard Cryo Freezer Box - for 15mL Tubes, 49 Places with Divider, 36/cs
SDP
Catalog No. NC1645152
Encompass
These sturdy cardboard boxes are specially coated to resist softening from repeated freeze and thaw cycles. Smart looking with a well sealing lid and a generous amount of room to write on top.
Catalog No. NC1645152 Supplier Stellar Scientific Supplier No. STO01536
These sturdy cardboard boxes are specially coated to resist softening from repeated freeze and thaw cycles. Smart looking with a well sealing lid and a generous amount of room to write on top.