Learn More
Research Products International Corp Mr. Frosty Cryo Freezing Container
SDP
Catalog No. 501977935
Encompass
The Mr. Frosty™ Cryo Freezing Container is excellent for laboratories with cryo preservation requirements. It provides the critical, repeatable -1°C per minute cooling rate required for successful cell preservation and recovery. Requires only isopropyl alcohol and a mechanical freezer. Convenient step-by-step directions are printed on the container. All components withstand repeated freezing/thawing cycles. Containers with alcohol can be stored at room temperature. Holds up to eighteen cryogenic vials.
Catalog No. 50-197-7935 Supplier Research Products International Corp Supplier No. 51000001
Add to Cart
Edge
Add to Cart
The Mr. Frosty™ Cryo Freezing Container is excellent for laboratories with cryo preservation requirements. It provides the critical, repeatable -1°C per minute cooling rate required for successful cell preservation and recovery. Requires only isopropyl alcohol and a mechanical freezer. Convenient step-by-step directions are printed on the container. All components withstand repeated freezing/thawing cycles. Containers with alcohol can be stored at room temperature. Holds up to eighteen cryogenic vials.